Dovitinib lactate 95%

Supplier: Ambeed
Warning

Synonyms: CHIR-258, TKI-258, 4-Amino-5-fluoro-3-(6-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)quinolin-2(1H)-one 2-hydroxypropanoate

A445850-10MG A445850-25MG A445850-50MG A445850-5MG A445850-1MG
76818-272EA 108.22 USD
76818-272 76818-274 76818-276 76818-278 77613-372
Dovitinib lactate 95%
Dovitinib lactate

Precaución: for Research Use Only

Formula: C₂₁H₂₁FN₆O·C₃H₆O₃
MW: 482.507 g/mol
MDL Number: MFCD16038905
CAS Number: 692737-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR