(2-Methylbenzo[d]oxazol-6-yl)boronic acid 98%

Supplier: Ambeed
Warning

A442988-1G A442988-250MG A442988-5G
76818-256EA 209 USD
76818-256 76818-258 76818-260
(2-Methylbenzo[d]oxazol-6-yl)boronic acid 98%
(2-Methylbenzo[d]oxazol-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.96 g/mol
MDL Number: MFCD10697425
CAS Number: 866332-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR