(1S,2S)-2-Aminocyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A441945-100MG A441945-250MG
76818-250EA 72.15 USD
76818-250 76818-252
(1S,2S)-2-Aminocyclopentanecarboxylic acid 95%
(1S,2S)-2-Aminocyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD09749904
CAS Number: 64191-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR