N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: N-[2-(Boc-amino)ethyl]glycine Ethyl Ester, Ethyl 2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate

A439229-1G A439229-5G A439229-25G
76818-234EA 38.2 USD
76818-234 76818-236 77673-076
N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester 98%
N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
MDL Number: MFCD00672310
CAS Number: 72648-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR