(5-Fluoro-2-iodophenyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-iodobenzyl alcohol

A437053-100MG A437053-1G A437053-250MG
76818-208EA 42.44 USD
76818-208 76818-210 76818-212
(5-Fluoro-2-iodophenyl)methanol 95%
(5-Fluoro-2-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆FIO
MW: 252.0248 g/mol
MDL Number: MFCD12031763
CAS Number: 877264-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR