3-(1-Pyrrolidinylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

A436028-1G A436028-250MG A436028-5G
76818-194EA 83.82 USD
76818-194 76818-196 76818-198
3-(1-Pyrrolidinylsulfonyl)benzeneboronic acid 98%
3-(1-Pyrrolidinylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₄S
MW: 255.10 g/mol
MDL Number: MFCD07783851
CAS Number: 871329-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR