2-Chloro-6-(trifluoromethyl)pyrazine 98%

Supplier: Ambeed
Warning

A435157-100MG A435157-1G A435157-250MG A435157-5G
76818-178EA 50.93 USD
76818-178 76818-180 76818-182 76818-184
2-Chloro-6-(trifluoromethyl)pyrazine 98%
2-Chloro-6-(trifluoromethyl)pyrazine

Precaución: for Research Use Only

Formula: C₅H₂ClF₃N₂
MDL Number: MFCD13189477
CAS Number: 61655-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR