2-Chloro-6-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-o-toluic acid

A433131-10G A433131-1G A433131-250MG A433131-25G A433131-5G
76818-164EA 77.01 USD
76818-164 76818-166 76818-168 76818-170 76818-172
2-Chloro-6-(trifluoromethyl)benzoic acid 98%
2-Chloro-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD06208254
CAS Number: 2376-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR