2-Amino-4-fluorobenzamide 98%

Supplier: Ambeed
Warning

A431725-1G A431725-250MG
76818-160EA 57 USD
76818-160 76818-162
2-Amino-4-fluorobenzamide 98%
2-Amino-4-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇FN₂O
MDL Number: MFCD11505061
CAS Number: 119023-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR