1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine] 97%

Supplier: Ambeed
Warning

Synonyms: Spiro[indole-3,4'-piperidin]-2(1H)-one, N1'-BOC protected, 1'-Boc-1,2-dihydro-2-oxo-spiro[3H-indole-3,4'-piperidine]

A431167-100MG A431167-1G A431167-250MG A431167-5G
76818-148EA 21.22 USD
76818-148 76818-150 76818-152 77594-338
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine] 97%
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine]

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₃
MW: 302.3682 g/mol
MDL Number: MFCD10566051
CAS Number: 252882-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR