N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine ≥97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-4-nitro-D-phenylalanine, Boc-D-Phe(4-NO2)-OH, Boc-D-Phe(4-NO₂)-OH

A431086-100G A431086-10G A431086-1G A431086-25G
76818-140EA 161.26 USD
76818-140 76818-142 76818-144 76818-146
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine ≥97%
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
MDL Number: MFCD00069880
CAS Number: 61280-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR