Ethyl 5-iodo-2-methylbenzoate 95+%

Supplier: Ambeed
Warning

A430875-100MG A430875-1G A430875-250MG A430875-5G
76818-132EA 10.61 USD
76818-132 76818-134 76818-136 76818-138
Ethyl 5-iodo-2-methylbenzoate 95+%
Ethyl 5-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₂
MW: 290.09 g/mol
MDL Number: MFCD11112849
CAS Number: 612833-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR