(3R,5S)-5-(Hydroxymethyl)-1-methylpyrrolidin-3-ol hydrochloride 98%

Supplier: Ambeed
Warning

A430604-1G A430604-250MG
76818-124EA 32 USD
76818-124 76818-126
(3R,5S)-5-(Hydroxymethyl)-1-methylpyrrolidin-3-ol hydrochloride 98%
(3R,5S)-5-(Hydroxymethyl)-1-methylpyrrolidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
MDL Number: MFCD12545916
CAS Number: 107746-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR