4-Chloro-(3-ethoxycarbonyl)phenylboronic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-3-(ethoxycarbony)phenylboronic acid, (4-Chloro-3-(ethoxycarbonyl)phenyl)boronic acid
A429740-10G
A429740-1G
A429740-250MG
A429740-5G
76818-098EA
13.8
USD
76818-098
76818-100
76818-102
76818-104
76818-106
4-Chloro-(3-ethoxycarbonyl)phenylboronic acid 96%
4-Chloro-(3-ethoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀BClO₄ |
MDL Number:
MFCD08235062 CAS Number: 874219-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...