7-Methyl-1H-pyrrolo[3,2-b]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A428556-100MG A428556-250MG
76818-094EA 102.91 USD
76818-094 76818-096
7-Methyl-1H-pyrrolo[3,2-b]pyridin-2(3H)-one 95%
7-Methyl-1H-pyrrolo[3,2-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.16 g/mol
MDL Number: MFCD12924242
CAS Number: 56057-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR