2-(1-(tert-Butoxycarbonyl)-4-hydroxypiperidin-4-yl)acetic acid 98%

Supplier: Ambeed
Warning

A427410-1G A427410-250MG A427410-100MG
76818-078EA 118 USD
76818-078 76818-080 77884-104
2-(1-(tert-Butoxycarbonyl)-4-hydroxypiperidin-4-yl)acetic acid 98%
2-(1-(tert-Butoxycarbonyl)-4-hydroxypiperidin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MDL Number: MFCD21090571
CAS Number: 502482-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR