(5-Bromopentyl)boronic acid 95%

Supplier: Ambeed
Warning

A426698-100MG A426698-1G A426698-250MG A426698-5G
76818-072EA 53 USD
76818-072 76818-074 76818-076 77884-100
(5-Bromopentyl)boronic acid 95%
(5-Bromopentyl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₁₂BBrO₂
MDL Number: MFCD10000909
CAS Number: 120986-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR