tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate 97%

Supplier: Ambeed
Warning

A426469-100MG A426469-1G A426469-250MG
76818-068EA 44 USD
76818-068 76818-070 77594-322
tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate 97%
tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD11858159
CAS Number: 1118786-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR