(3,5-Difluoro-4-(methoxycarbonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-4-methoxycarbonylphenylboronic acid

A425854-100MG A425854-250MG A425854-5G A425854-1G
76818-062EA 34 USD
76818-062 76818-064 76818-066 77594-318
(3,5-Difluoro-4-(methoxycarbonyl)phenyl)boronic acid 98%
(3,5-Difluoro-4-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₂O₄
MW: 215.95 g/mol
MDL Number: MFCD16295262
CAS Number: 1190989-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR