7-Bromo-6-methoxy-5-azaindole 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-6-methoxy-1H-pyrrolo[3,2-c]pyridine

A422521-100MG A422521-1G A422521-250MG
76818-050EA 37 USD
76818-050 76818-052 76818-054
7-Bromo-6-methoxy-5-azaindole 97%
7-Bromo-6-methoxy-5-azaindole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
MDL Number: MFCD12962570
CAS Number: 1190314-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR