4-Bromo-7-chloroisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A419422-1G A419422-250MG
76818-040EA 328 USD
76818-040 76818-042
4-Bromo-7-chloroisoquinolin-1(2H)-one 95%
4-Bromo-7-chloroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
MDL Number: MFCD13193317
CAS Number: 1028252-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR