Morpholin-3-ylmethanol hydrochloride 98%

Supplier: Ambeed
Warning

A417562-1G A417562-250MG A417562-5G
76818-034EA 57.29 USD
76818-034 76818-036 76818-038
Morpholin-3-ylmethanol hydrochloride 98%
Morpholin-3-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·ClH
MW: 153.607 g/mol
MDL Number: MFCD12547467
CAS Number: 955400-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR