Azetidin-3-amine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Aminoazetidine dihydrochloride
A416938-250MG
A416938-5G
76818-024EA
22.28
USD
76818-024
76818-026
Azetidin-3-amine dihydrochloride 97%
Azetidin-3-amine dihydrochloride
Precaución: for Research Use Only
Formula:
C₃H₁₀Cl₂N₂ MW: 145.03 g/mol |
MDL Number:
MFCD09910173 CAS Number: 102065-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...