1-Cyclopropyl-4-piperidinone ≥98%

Supplier: Ambeed
Danger

A416757-25G A416757-5G
76818-022EA 9.55 USD
76818-022 77884-082 77884-083
1-Cyclopropyl-4-piperidinone ≥98%
1-Cyclopropyl-4-piperidinone

Precaución: for Research Use Only

Formula: C₈H₁₃NO
MDL Number: MFCD07374375
CAS Number: 62813-01-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR