ω-Bromo-4-chloroacetophenone 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Bromo-4'-chloroacetophenone, 2-Bromo-1-(4-chlorophenyl)ethanone, Bromomethyl p-chlorophenyl ketone, α-Bromo-4-chloroacetophenone, 2-Bromo-4′-chloroacetophenone, 4-Chlorophenacyl bromide
A416000-10G
A416000-25G
A416000-5G
A416000-100G
76818-016EA
22
USD
76818-016
76818-018
76818-020
77884-079
ω-Bromo-4-chloroacetophenone 97%
ω-Bromo-4-chloroacetophenone
Precaución: for Research Use Only
Formula:
C₈H₆BrClO |
MDL Number:
MFCD00000625 CAS Number: 536-38-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...