Alizarine 95%

Supplier: Ambeed
Warning

Synonyms: Alizarin, 1,2-Dihydroxy-9,10-anthraquinone, 1,2-Dihydroxy-9,10-anthracenedione, 1,2-Dihydroxyanthraquinone

A415687-100MG A415687-25G A415687-100G
76818-014EA 6 USD
76818-014 77568-842 77884-076
Alizarine 95%
Alizarine

Precaución: For research use only.

Formula: C₁₄H₈O₄
MDL Number: MFCD00001201
CAS Number: 72-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR