(3-Acetamido-4-fluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A415393-100MG A415393-1G A415393-250MG
76818-004EA 56 USD
76818-004 76818-006 76818-008
(3-Acetamido-4-fluorophenyl)boronic acid 98%
(3-Acetamido-4-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
MW: 196.97 g/mol
MDL Number: MFCD21362227
CAS Number: 1426255-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR