Methyl-5-bromoquinoline-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromoquinoline-2-carboxylate

A413951-100MG A413951-1G A413951-250MG
76817-996EA 38 USD
76817-996 76817-998 76818-000
Methyl-5-bromoquinoline-2-carboxylate 97%
Methyl 5-bromoquinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO₂
MDL Number: MFCD21086458
CAS Number: 1355174-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR