2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Nitro-6-(trifluoromethyl)pyridin-2(1H)-one
A413147-100MG
A413147-1G
A413147-250MG
A413147-25G
A413147-5G
76817-986EA
7
USD
76817-986
76817-988
76817-990
76817-992
76817-994
2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine 98%
2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₃F₃N₂O₃ MW: 208.1 g/mol |
MDL Number:
MFCD10696218 CAS Number: 117519-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...