6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1-methyl-3,4-dihydroquinolin-2(1H)-one, 6-Bromo-1-methyl-2-oxo-1,2,3,4-tetrahydroquinoline

A409445-1G A409445-250MG A409445-5G
76817-964EA 62.6 USD
76817-964 76817-966 76817-968
6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one 95%
6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.097 g/mol
MDL Number: MFCD18266837
CAS Number: 1092523-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR