4-(4-Bromophenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Bromophenyl)carbamoyl)phenyl)boronic acid, 4-(4-Bromophenylcarbamoyl)benzeneboronic acid

A409417-1G A409417-25G A409417-5G
76817-958EA 11 USD
76817-958 76817-960 76817-962
4-(4-Bromophenylcarbamoyl)phenylboronic acid 98%
4-(4-Bromophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BBrNO₃
MDL Number: MFCD09027201
CAS Number: 874287-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR