1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine 95%

Supplier: Ambeed
Warning

A409037-100MG A409037-1G A409037-250MG
76817-952EA 102.91 USD
76817-952 76817-954 76817-956
1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine 95%
1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₅H₆F₃N₃
MW: 165.11 g/mol
MDL Number: MFCD03420215
CAS Number: 919278-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR