(1H-Pyrazol-5-yl)boronic acid 97%

Supplier: Ambeed
Warning

A407336-1G A407336-250MG A407336-5G
76817-944EA 49.87 USD
76817-944 76817-946 76817-948
(1H-Pyrazol-5-yl)boronic acid 97%
(1H-Pyrazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₃H₅BN₂O₂
MW: 111.89 g/mol
MDL Number: MFCD07368247
CAS Number: 724710-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR