t-Butyl (3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-isopropylidenedioxy-6-heptenoate 95%

Supplier: Ambeed
Warning

A405187-5G A405187-1G A405187-250MG
76817-932EA 23 USD
76817-932 77613-240 77613-242
t-Butyl (3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-isopropylidenedioxy-6-heptenoate 95%
t-Butyl (3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-isopropylidenedioxy-6-heptenoate

Precaución: for Research Use Only

Formula: C₃₂H₃₆FNO₄
MDL Number: MFCD12911897
CAS Number: 147489-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR