7,10-Dichloro-2-methoxybenzo[b]-1,5-naphthyridine 95%

Supplier: Ambeed
Warning

A404706-250MG A404706-5G A404706-1G
76817-928EA 13 USD
76817-928 76817-930 77884-054
7,10-Dichloro-2-methoxybenzo[b]-1,5-naphthyridine 95%
7,10-Dichloro-2-methoxybenzo[b]-1,5-naphthyridine

Precaución: for Research Use Only

Formula: C₁₃H₈Cl₂N₂O
MDL Number: MFCD00160720
CAS Number: 6626-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR