(3-(Benzyloxy)-4-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzyloxy)-4-methylphenylboronic acid

A404642-1G A404642-5G
76817-924EA 44.56 USD
76817-924 76817-926
(3-(Benzyloxy)-4-methylphenyl)boronic acid 98%
(3-(Benzyloxy)-4-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₃
MW: 242.08 g/mol
MDL Number: MFCD11617255
CAS Number: 1256355-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR