4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-[(N-methoxy-N-methyl)carbamoyl]phenylboronic acid, (4-Fluoro-3-(methoxy(methyl)carbamoyl)phenyl)boronic acid, 4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid

A403296-1G A403296-250MG A403296-5G
76817-902EA 50 USD
76817-902 76817-904 76817-906
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₄
MDL Number: MFCD08235106
CAS Number: 874290-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR