4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride 95%

Supplier: Ambeed
Warning

A402932-100MG A402932-10G A402932-1G A402932-250MG A402932-5G A402932-25G A402932-100G
76817-892EA 5 USD
76817-892 76817-894 76817-896 76817-898 76817-900 77672-970 77884-053
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride 95%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁Cl₂N₃
MDL Number: MFCD00182087
CAS Number: 62002-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR