(5-Formyl-2-methylphenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A401980-100MG A401980-1G A401980-250MG
76817-886EA 33 USD
76817-886 76817-888 76817-890
(5-Formyl-2-methylphenyl)boronic acid 95+%
(5-Formyl-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MDL Number: MFCD16295249
CAS Number: 1186398-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR