(3,3-Difluorocyclobutyl)methanol 97%

Supplier: Ambeed
Danger

A401673-100MG A401673-1G A401673-250MG A401673-25G A401673-5G
76817-872EA 7.43 USD
76817-872 76817-874 76817-876 76817-878 76817-880
(3,3-Difluorocyclobutyl)methanol 97%
(3,3-Difluorocyclobutyl)methanol

Precaución: for Research Use Only

Formula: C₅H₈F₂O
MDL Number: MFCD11877861
CAS Number: 681128-39-2
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR