6-Bromo-1H-pyrrolo[3,2-c]pyridine 98%

Supplier: Ambeed
Warning

A398684-100MG A398684-1G A398684-250MG A398684-5G
76817-864EA 37.14 USD
76817-864 76817-866 76817-868 76817-870
6-Bromo-1H-pyrrolo[3,2-c]pyridine 98%
6-Bromo-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD08690134
CAS Number: 1000342-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR