1,3-Didodecyl-2-methyl-1H-imidazol-3-ium chloride 95%

Supplier: Ambeed
Warning

A398680-100G A398680-1G A398680-25G A398680-5G
76817-856EA 213.25 USD
76817-856 76817-858 76817-860 76817-862
1,3-Didodecyl-2-methyl-1H-imidazol-3-ium chloride 95%
1,3-Didodecyl-2-methyl-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₂₈H₅₅ClN₂
MW: 455.20 g/mol
MDL Number: MFCD11617810
CAS Number: 21054-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR