2-Methoxy-6-methylpyridine 98%

Supplier: Ambeed
Danger

A398419-10G A398419-25G A398419-5G A398419-100G
76817-852EA 10 USD
76817-852 76817-854 77672-964 77884-040
2-Methoxy-6-methylpyridine 98%
2-Methoxy-6-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD04112507
CAS Number: 63071-03-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR