6-Chloro-5-methylnicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-3-methylpyridine

A395641-100MG A395641-1G A395641-250MG A395641-5G
76817-838EA 55.17 USD
76817-838 76817-840 76817-842 76817-844
6-Chloro-5-methylnicotinic acid 97%
6-Chloro-5-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
MDL Number: MFCD17214192
CAS Number: 66909-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR