(3R,4S,5S)-Tetrahydro-2H-pyran-2,3,4,5-tetraol 98%

Supplier: Ambeed
Warning

A395369-1000G A395369-100G A395369-100MG A395369-10G A395369-25G A395369-500G A395369-5G
76817-822EA 111.4 USD
76817-822 76817-824 76817-826 76817-828 76817-830 76817-832 76817-834
(3R,4S,5S)-Tetrahydro-2H-pyran-2,3,4,5-tetraol 98%
(3R,4S,5S)-Tetrahydro-2H-pyran-2,3,4,5-tetraol

Precaución: for Research Use Only

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00006609
CAS Number: 87-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR