3-Chloro-4-(methylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(N-methylcarbamoyl)phenylboronic acid, (3-Chloro-4-(methylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(methylcarbamoyl)benzeneboronic acid

A392658-100MG A392658-1G A392658-250MG
76817-784EA 8 USD
76817-784 76817-786 76817-788
3-Chloro-4-(methylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BClNO₃
MDL Number: MFCD07363766
CAS Number: 850589-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR