1-(6-Methylpyridin-3-yl)ethanamine 95%

Supplier: Ambeed
Danger

A392625-1G A392625-250MG A392625-100MG
76817-780EA 445.58 USD
76817-780 76817-782 77672-948
1-(6-Methylpyridin-3-yl)ethanamine 95%
1-(6-Methylpyridin-3-yl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.19 g/mol
MDL Number: MFCD09864336
CAS Number: 92295-43-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR