5-Bromo-2-chloro-4-fluoroaniline 95%

Supplier: Ambeed
Warning

A389149-250MG A389149-1G A389149-5G
76817-768EA 9 USD
76817-768 77672-936 77884-030
5-Bromo-2-chloro-4-fluoroaniline 95%
5-Bromo-2-chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD21603962
CAS Number: 305795-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR