3-Methylpiperidine-3-carboxylic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A389003-1G
A389003-250MG
A389003-25G
A389003-5G
76817-760EA
34
USD
76817-760
76817-762
76817-764
76817-766
3-Methylpiperidine-3-carboxylic acid hydrochloride 98%
3-Methylpiperidine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD20441739 CAS Number: 116140-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...