Dimethyl-1H-indole-2,6-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dimethyl 1H-indole-2,6-dicarboxylate
A387496-1G
A387496-250MG
76817-744EA
61
USD
76817-744
76817-746
Dimethyl-1H-indole-2,6-dicarboxylate 97%
Dimethyl-1H-indole-2,6-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₄ |
MDL Number:
MFCD06653349 CAS Number: 881040-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...